Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRAS Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299921DL594
Description
HRAS Polyclonal specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA.Specifications
HRAS | |
Polyclonal | |
Western Blot, ELISA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
DyLight 594 | |
50mM Sodium Borate | |
Produced in rabbits immunized with purified, recombinant HRAS (P01112; Met1-Cys186). | |
0.1 mL | |
Cell Cycle and Replication, Metastasis, mTOR Pathway | |
3265 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction