Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRAS Antibody (H-Ras-03), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068799AF350
Description
HRAS Monoclonal specifically detects HRAS in Human samples. It is validated for Western Blot.Specifications
HRAS | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
3265 | |
Human | |
IgG1 |
Western Blot | |
H-Ras-03 | |
Western Blot | |
HRAS | |
Peptide corresponding to amino acids DIHQYREQIKRVKDSDDC of human H-Ras protein. | |
Primary | |
The antibody H-RAS-03 reacts with human H-Ras, an ubiquitously expressed 21 kDa intracellular protein. Although reactivity with other species has not been determined, it is probable as the epitope is highly conserved among animals. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction