Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRAS Antibody (H-Ras-03) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11068799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11068799 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11068799 Supplier Novus Biologicals Supplier No. NB11068799
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HRAS Monoclonal specifically detects HRAS in Human samples. It is validated for Western Blot.

Specifications

Antigen vH RAS
Applications Western Blot
Classification Monoclonal
Clone H-Ras-03
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 -1 μg/mL
Gene Accession No. P01112
Gene Alias C-BAS/HAS, C-HA-RAS1, c-has/bas p21 protein, C-H-RAS, c-ras-Ki-2 activated oncogene, CTLO, GTP- and GDP-binding peptide B, GTPase HRas, HAMSV, Ha-Ras, Ha-Ras1 proto-oncoprotein, H-Ras-1, HRAS1, H-RASIDX, K-RAS, N-RAS, p19 H-RasIDX protein, p21ras, Ras family small GTP binding protein H-Ras, RASH1, transformation gene: oncogene HAMSV, Transforming protein p21, v-Ha-ras Harvey rat sarcoma viral oncogene homolog
Gene Symbols HRAS
Host Species Mouse
Immunogen Peptide corresponding to amino acids DIHQYREQIKRVKDSDDC of human H-Ras protein.
Molecular Weight of Antigen 21 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3265
Test Specificity The antibody H-RAS-03 reacts with human H-Ras, an ubiquitously expressed 21 kDa intracellular protein. Although reactivity with other species has not been determined, it is probable as the epitope is highly conserved among animals.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.