Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRAS Antibody (OTI1D9), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274847H
Description
HRAS Monoclonal specifically detects HRAS in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HRAS | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Metastasis, mTOR Pathway | |
3265.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1D9 | |
Western Blot, Immunohistochemistry | |
HRAS | |
Full length recombinant protein of human vH RAS (NP_005334) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction