Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRAS Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299921PE
Description
HRAS Polyclonal antibody specifically detects HRAS in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
HRAS | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
Produced in rabbits immunized with purified, recombinant Human HRAS (Accession#: P01112; Met1-Cys186) | |
0.1 mL | |
Cell Cycle and Replication, Metastasis, mTOR Pathway | |
3265 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction