Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRAS like suppressor Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10093406
Description
HRAS like suppressor Polyclonal specifically detects HRAS like suppressor in Human, Mouse, Rat samples. It is validated for Peptide ELISA.Specifications
HRAS like suppressor | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
A-C1, HRAS-like suppressor, HRASLS1, H-REV107, H-REV107 protein-related protein, HSD28, phospholipase A | |
Goat | |
Affinity Purified | |
RUO | |
57110 | |
Store at -20C. Avoid freeze-thaw cycles. |
ELISA | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:2000 | |
NP_065119.1 | |
PLAAT1 | |
Peptide with sequence C-RYGEGVSEQANR corresponding to internal region according to NP_065119.1. | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction