Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRAS Rat anti-Human, Rat, Clone: Y13-259, Abnova™

Catalog No. 89035588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-588 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-588 Supplier Abnova Corporation Supplier No. MAB14191

Rat Monoclonal Antibody

Rat monoclonal antibody raised against recombinant human HRAS.

  • High specificity
  • High reactivity

Specifications

Antigen HRAS
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Y13-259
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant human HRAS.
Dilution Immunofluorescence Immunohistochemistry Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. P01112
Gene Alias C-BAS/HAS/C-H-RAS/C-HA-RAS1/CTLO/H-RASIDX/HAMSV/HRAS1/K-RAS/N-RAS/RASH1
Gene Symbols HRAS
Host Species Rat
Immunogen Recombinant protein corresponding to human HRAS.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3265
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.