Learn More
Invitrogen™ HRASLS Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000. Preliminary analysis showed no signal in Western Blot.
Specifications
Specifications
| Antigen | HRASLS |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | HRASLS |
| Gene Accession No. | Q9HDD0 |
| Gene Alias | A-C1; HRAS like suppressor; HRAS-like suppressor 1; HRASLS; HRASLS1; H-REV107; H-REV107 protein-related protein; HRSL1; HSD28; phospholipase A; Phospholipase A and acyltransferase 1; Phospholipid-metabolizing enzyme A-C1; PLAAT1 |
| Gene Symbols | HRASLS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.