Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRASLS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563640
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563640 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563640 Supplier Invitrogen™ Supplier No. PA563640
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AIVKKELLSV VAGGDNYRVN NKHDD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.

HRASLS2 (HRAS-like suppressor 2), also known as PLA1/2-2, is a 162 amino acid Cytoplasmic protein that belongs to the H-rev107 family. Expressed in liver, kidney, small intestine and colon, HRASLS2 exhibits PLA1/2 activity by catalyzing the calcium-independent hydrolysis of acyl groups in various phosphotidylcholines (PCs) and phosphatidylethanolamine (PE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRASLS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HRASLS2
Gene Accession No. Q9NWW9
Gene Alias HRAS like suppressor 2; HRAS-like suppressor 2; HRASLS2; Phospholipase A and acyltransferase 2; PLA1/2-2; PLAAT2
Gene Symbols HRASLS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HRASLS2. Recombinant protein control fragment (Product #RP-101923).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.