Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRASLS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145228 Supplier Invitrogen™ Supplier No. PA5145228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVQLPAKQPP PGTLEQGRSI QQGEKAVVSL ETTPSQKADW SSIPKPENEG KLIKQAAEGK PRPRPG Antigen sequence identity: Mouse 50%; Rat 38%.

Exhibits both phospholipase A1/2 and acyltransferase activities (PubMed:22825852, PubMed:26503625). Shows phospholipase A1 (PLA1) and A2 (PLA2) activity, catalyzing the calcium-independent release of fatty acids from the sn-1 or sn-2 position of glycerophospholipids (PubMed:22825852). Shows N-acyltransferase activity, catalyzing the calcium-independent transfer of a fatty acyl group at the sn-1 position of phosphatidylcholine (PC) and other glycerophospholipids to the primary amine of phosphatidylethanolamine (PE), forming N-acylphosphatidylethanolamine (NAPE), which serves as precursor for N-acylethanolamines (NAEs) (PubMed:19000777, PubMed:22825852). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRASLS5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HRASLS5
Gene Accession No. Q96KN8
Gene Alias 1700055A14Rik; 1700086E08Rik; 4921526K24Rik; Ca(2+)-independent N-acyltransferase; Ca-independent N-acyltransferase; calcium-independent phosphatidylethanolamine N-acyltransferase; HRAS like suppressor family member 5; HRAS-like suppressor 5; HRAS-like suppressor family, member 5; Hrasls5; H-rev107-like protein 5; HRLP5; HRSL5; iNAT; lecithin-retinol acyltransferase (LRAT)-like protein-1; Phospholipase A and acyltransferase 5; PLAAT5; RGD1308376; RLP1; testicular tissue protein Li 90
Gene Symbols HRASLS5
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 68-133. Recombinant protein control fragment (Product #RP-109896).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.