Learn More
Invitrogen™ HRD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596185
Description
Positive Samples: HeLa, HepG2, Mouse pancreas, Mouse lung, Mouse kidney; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein Immunogen sequence: LEARLQSLRN IHTLLDAAML QINQYLTVLA SLGPPRPATS VNSTEETATT VVAAASSTSI PSSEATTPTP GASPPAPEME RPPAPESVGT EEMPEDGEPD A.
HRD1 is a ubiquitin ligase whose expression is induced by the unfolded protein response (UPR) following endoplasmic reticulum stress. Expression of HRD1 protects cells from apoptosis by inducing degradation of abnormally processed proteins that accumulate in the endoplasmic reticulum. HRD1 is expressed in many tissues, strongly expressed in brain, pancreas, liver, kidney and skeletal muscle. It has been reported that Synoviolin/Hrd1 (expressed in rheumatoid synovium) is a novel causative factor for arthropathy by triggering synovial cell outgrowth through its anti-apoptotic effects. HRD1 contains one ring-type zinc finger.
Specifications
HRD1 | |
Polyclonal | |
Unconjugated | |
SYVN1 | |
1200010C09Rik; AW211966; C85322; D530017H19Rik; DER3; E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; Hrd1; HRD1 protein; KIAA1810; MGC40372; RGD1310488; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin; synoviolin 1; Syvn1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
74126, 84447 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q86TM6, Q9DBY1 | |
SYVN1 | |
A synthetic peptide corresponding to a sequence within amino acids 500-600 of human SYVN1 (NP_7573851). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.