Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA146121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA146121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA146121 Supplier Invitrogen™ Supplier No. PA146121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: 293T, INS-1, and MIN6 cell lysates, antigen standard for SYVN1 (transient overexpression lysate).

HRD1 is a ubiquitin ligase whose expression is induced by the unfolded protein response (UPR) following endoplasmic reticulum stress. Expression of HRD1 protects cells from apoptosis by inducing degradation of abnormally processed proteins that accumulate in the endoplasmic reticulum. HRD1 is expressed in many tissues, strongly expressed in brain, pancreas, liver, kidney and skeletal muscle. It has been reported that Synoviolin/Hrd1 (expressed in rheumatoid synovium) is a novel causative factor for arthropathy by triggering synovial cell outgrowth through its anti-apoptotic effects. HRD1 contains one ring-type zinc finger.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SYVN1
Gene Accession No. Q86TM6, Q9DBY1
Gene Alias 1200010C09Rik; AW211966; C85322; D530017H19Rik; DER3; E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; Hrd1; HRD1 protein; KIAA1810; MGC40372; RGD1310488; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin; synoviolin 1; Syvn1
Gene Symbols SYVN1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human HRD1 (within residues 300-400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100765313, 74126, 84447
Target Species Hamster, Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.