Learn More
Description
Specifications
Specifications
| Antigen | HRD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 155mM NaCl, 50mM Tris-HCl pH 7.4 with 0.05% Sodium Azide and 10% glycerol. |
| Gene | SYVN1, HRD1, KIAA1810 |
| Gene Accession No. | Q86TM6 |
| Host Species | Rabbit |
| Immunogen | Linear peptide corresponding to the C-terminal sequence of human HRD1. |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.