Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRG Monoclonal Antibody (C3), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $471.00
Specifications
Antigen | HRG |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550999
|
Invitrogen™
MA550999 |
200 μL |
Each for $471.00
|
|
|||||
FEMA551000
|
Invitrogen™
MA551000 |
20 μL |
Each for $195.80
|
|
|||||
Description
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.Specifications
HRG | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11 | |
HRG | |
IgG2b κ | |
Protein A/G | |
HRG |
C3 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
Q99PS8 | |
171016 | |
Recombinant Eukaryotic Rat HRG protein, Leu19-Lys525 (Accession #Q99PS8). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title