Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597051
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597051 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597051 Supplier Invitrogen™ Supplier No. PA597051
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPTDCSAVEP EAEKALDLIN KRRRDGYLFQ LLRIADAHLD RVENTTVYYL VLDVQESDCS VLSRKYWNDC EPPDSRRPSE IVIGQCKVIA TRHSHESQDL RVIDFNCTTS SVSSALANTK DSPVLIDFFE DTERYRKQAN KALEKYKEEN DDFASFRVDR IERVARVRGG EGTGYFVDFS VRNCPRHHFP RHPNVFGFCR ADLFYDVEAL DLESPKNLVI NCEVFDPQEH ENINGVPPHL G; Positive Samples: Mouse pancreas.

HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRG
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HRG
Gene Accession No. Q99PS8, Q9ESB3
Gene Alias AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11
Gene Symbols HRG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-260 of human HRG (NP_0004031).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171016, 94175
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.