Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120691 Supplier Invitrogen™ Supplier No. PA5120691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-251MG, Mouse liver, Rat liver. Immunogen sequence: KPGKESPWEV LKRKPKDAGG GSVLKSPSQT PKEMKSPVVF SQEDDREVDK LYCFPLDIVH MQAAAEGSSR DYVAVNRSHG QLKTDEQGLN T.

Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Its various actions are mediated by histamine receptors H1, H2, H3 and H4. This gene was thought to be intronless until recently. The protein encoded by this gene is an integral membrane protein and belongs to the G protein-coupled receptor superfamily. It mediates the contraction of smooth muscles, the increase in capillary permeability due to contraction of terminal venules, the release of catecholamine from adrenal medulla, and neurotransmission in the central nervous system. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRH1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HRH1
Gene Accession No. P31390, P35367, P70174
Gene Alias Bordetella pertussis induced histamine sensitization; Bphs; H1 receptor; H1R; H1-R; HH1R; Hir; hisH1; Hisr; histamine H1 receptor; Histamine H1-R; histamine receptor; histamine receptor H 1; histamine receptor H1; histamine receptor, subclass H1; Hrh1
Gene Symbols HRH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-340 of human HRH1 (NP_000852.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15465, 24448, 3269
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.