Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ HRH2 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142779
                        
                            
                        
                        
                        
                            
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001124527.1; NP_071640.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
H2 Histamine Receptor stimulates adenylate cyclase activity and leads to the release of acids in the stomach. It is also involved in GI motility and intestinal secretion. Treatments for acid-peptic disorders are based on the inhibition of histamine H2 receptor activity. The 3-untranslated region may be involved in differential expression. Histamine H2 receptor has been reported to be expressed in bone marrow, brain, heart, lymph node, nose, placenta, spinal cord, and stomach. An EST has been isolated from a kidney library. 
            Specifications
| HRH2 | |
| Polyclonal | |
| Unconjugated | |
| HRH2 | |
| gastric receptor 1; gastric receptor I; H2R; HH2R; histamine H2 receptor; histamine receptor; histamine receptor H 2; histamine receptor H2; HRH2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 15466, 25461 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P25102, P97292 | |
| HRH2 | |
| Peptide with sequence C-SHNSHKTSLRLNNS (aa 308-321). | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            