Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRH3 Recombinant Rabbit Monoclonal Antibody (27H8)
GREENER_CHOICE

Catalog No. FEMA550181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550181 Supplier Invitrogen™ Supplier No. MA550181
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HRH3 Recombinant Monoclonal Antibody for Flow, ELISA

H3 Histamine Receptor regulates the release of the neurotransmitters norepinephrine and somatostatin. In histamine-containing neurons of the brain, the H3 receptor acts as a presynaptic autoreceptor, while in nonhistamine-containing neurons, in both the central and peripheral nervous systems, it acts as a presynaptic heteroreceptor. Multiple isoforms of H3 receptors are produced by alternative splicing. Histamine H3 receptor has been reported primarily in brain. ESTs have been isolated from brain (normal and cancer), breast cancer, colon cancer, and retinoblastoma libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRH3
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 27H8
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HRH3
Gene Accession No. Q9Y5N1
Gene Alias AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3
Gene Symbols HRH3
Host Species Rabbit
Immunogen A synthesized peptide derived from Human HRH3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11255
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.