Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HRH3 Recombinant Rabbit Monoclonal Antibody (7H8E6)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538074
Description
Positive Samples: Mouse testis, Mouse brain, Mouse spleen, Rat testis, Rat brain, Rat spleen, Raji, SH-SY5Y, U-251MG Immunogen sequence: MERAPPDGPL NASGALAGEA AAAGGARGFS AAWTAVLAAL MALLIVATVL GNALVMLAFV ADSSLRTQNN FFLLNLAISD FLVGAFCIPL YVPYVLTGRW.
H3 Histamine Receptor regulates the release of the neurotransmitters norepinephrine and somatostatin. In histamine-containing neurons of the brain, the H3 receptor acts as a presynaptic autoreceptor, while in nonhistamine-containing neurons, in both the central and peripheral nervous systems, it acts as a presynaptic heteroreceptor. Multiple isoforms of H3 receptors are produced by alternative splicing. Histamine H3 receptor has been reported primarily in brain. ESTs have been isolated from brain (normal and cancer), breast cancer, colon cancer, and retinoblastoma libraries.
Specifications
| HRH3 | |
| Recombinant Monoclonal | |
| 0.74 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58406, Q9QYN8, Q9Y5N1 | |
| HRH3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HRH3 (Q9Y5N1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| 7H8E6 | |
| Unconjugated | |
| HRH3 | |
| AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11255, 85268, 99296 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction