Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589632 Supplier Invitrogen™ Supplier No. PA589632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-431, SH-SY5Y; Cellular Location: Cytoplasm Immunogen sequence: SIVEQQLPLR RNSHLEESFT STEESSEENV NFLGQTEAQY HLMLHIQMQL CELSLWDWIV ERNKRGREYV DESACPYVMA NVATKIFQEL VEGVFYIHNM GIVHRDLKPR NIFLHGPDQQ VKIGDFGLAC TDILQKNTDW TNRNGKRTPT HTSRVGTCLY ASPEQLEGSE YDAKSDMYSL GVVLLELFQP FGTEMERAEV LTGLRTGQLP ESLRKRCPVQ AKYIQHLTRR NSSQRPSAIQ LLQSELFQNS.

EIF2AK1 (HRI) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK1, signaling is initiated by low levels of heme. The HRI gene is localized to 7p22 where its 3' end slightly overlaps the 3' end of the gene JTV1. The two genes are transcribed from opposite strands and kinase activity is induced by low levels of heme and inhibited by the presence of heme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRI
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EIF2AK1
Gene Accession No. Q63185, Q9BQI3
Gene Alias EIF2AK1; eukaryotic translation initiation factor 2 alpha kinase 1; eukaryotic translation initiation factor 2-alpha kinase 1; HCR; he; heme regulated initiation factor 2 alpha kinase; heme sensitive initiation factor 2a kinase; heme-controlled repressor; Heme-regulated eukaryotic initiation factor eIF-2-alpha kinase; heme-regulated inhibitor; heme-regulated initiation factor 2-alpha kinase; heme-regulated repressor; hemin-sensitive initiation factor 2a kinase; hemin-sensitive initiation factor 2-alpha kinase; Hri; KIAA1369; PRO1362
Gene Symbols EIF2AK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 337-586 of human EIF2AK1 (NP_0552282).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27102, 27137
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.