Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBY1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549667
Description
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
CBY1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y3M2 | |
CBY1 | |
KLH conjugated synthetic peptide between 98-126 amino acids from the C-terminal region of human HRIHFB2025. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CBY1 | |
Protein chibby homolog 1, ARPP-binding protein, Cytosolic leucine-rich protein, PIGEA-14, PKD2 interactor, Golgi and endoplasmic reticulum-associated 1, CBY1, ARB1, C22orf2, CBY, PGEA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25776 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction