Learn More
Invitrogen™ HRK Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 83%; Rabbit: 100%; Rat: 83%.
Specifications
Specifications
| Antigen | HRK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | HRK |
| Gene Accession No. | O00198 |
| Gene Alias | Activator of apoptosis harakiri; activator of apoptosis Hrk; AI838259; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; Bid3; death protein 5; DP5; harakiri; harakiri, BCL2 interacting protein; harakiri, BCL2 interacting protein (contains only BH3 domain); Hrk; Neuronal death protein DP5 |
| Gene Symbols | HRK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.