Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRK Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA186773

Catalog No. PIPA186773


Only null left
Add to cart

Description

Description

In Western blot, this antibody detects a band at ~13 kDa in mouse pancreas tissue lysate. Rabbit anti HRK antibody detects Harakiri (HRK), also known as Neuronal death protein DP5 or BH3-interacting domain-containing protein 3. HRK is a BH3 only protein of the Bcl-2 family, which promotes Bax dependent apoptosis.

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Hrk, a pro-apoptotic member of the Bcl-2 homology domain-3 (BH3)-only group of the Bcl-2 family of proteins, was also identified as novel protein induced during programmed neuronal death. It lacks significant homology to other Bcl-2 family members except for an 8-amino acid region that is similar to the BH3 motif of Bik. Hrk regulates apoptosis through interaction with the anti-apoptotic proteins Bcl-2 and Bcl-XL via this domain. It does not interact with the pro-apoptotic proteins Bax, Bak, or Bcl-XS. Hrk localizes to mitochondrial membranes in a pattern similar to that previously reported for Bcl-2 and Bcl-XL.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HRK
Polyclonal
Unconjugated
HRK
Activator of apoptosis harakiri; activator of apoptosis Hrk; AI838259; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; Bid3; death protein 5; DP5; harakiri; harakiri, BCL2 interacting protein; harakiri, BCL2 interacting protein (contains only BH3 domain); Hrk; Neuronal death protein DP5
Rabbit
Affinity chromatography
RUO
117271, 12123, 8739
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O00198, P62816, P62817
HRK
15 amino acid peptide from near the centre of human HRK.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.