Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hrk Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10302R

Description
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Hrk, a pro-apoptotic member of the Bcl-2 homology domain-3 (BH3)-only group of the Bcl-2 family of proteins, was also identified as novel protein induced during programmed neuronal death. It lacks significant homology to other Bcl-2 family members except for an 8-amino acid region that is similar to the BH3 motif of Bik. Hrk regulates apoptosis through interaction with the anti-apoptotic proteins Bcl-2 and Bcl-XL via this domain. It does not interact with the pro-apoptotic proteins Bax, Bak, or Bcl-XS. Hrk localizes to mitochondrial membranes in a pattern similar to that previously reported for Bcl-2 and Bcl-XL.Specifications
Hrk | |
Polyclonal | |
Unconjugated | |
HRK | |
Activator of apoptosis harakiri; activator of apoptosis Hrk; AI838259; BCL2-interacting protein; BH3 interacting (with BCL2 family) domain, apoptosis agonist; BH3-interacting domain-containing protein 3; Bid3; death protein 5; Dp5; harakiri; harakiri, BCL2 interacting protein; harakiri, BCL2 interacting protein (contains only BH3 domain); HRK; Neuronal death protein DP5 | |
Rabbit | |
Protein A | |
RUO | |
8739 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O00198 | |
HRK | |
KLH conjugated synthetic peptide derived from human Activator of apoptosis harakiri. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction