Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hrk Rabbit anti-Human, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF851SP
This item is not returnable.
View return policy
Description
Hrk Polyclonal antibody AF851SP specifically detects Hrk in Human samples. It is validated for Western BlottingSpecifications
Hrk | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HRK | |
KLH-coupled human Hrk synthetic peptide, RTMWRRRARSR RAPAPGAALPC | |
25 μg | |
Primary | |
Detects human Hrk in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
activator of apoptosis harakiri, activator of apoptosis Hrk, BCL2-interacting protein, BH3-interacting domain-containing protein 3, BID3, death protein 5, DP5, HARAKIRI, harakiri, BCL2 interacting protein (contains only BH3 domain), harakiri, BCL2-interacting protein (contains only BH3 domain), Neuronal death protein DP5 | |
Rabbit | |
Affinity Purified | |
RUO | |
8739 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction