Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRP Antibody (HP-03), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500420AF700
Description
HRP Monoclonal specifically detects HRP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
HRP | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
The antibody HP-03 reacts with horseradish peroxidase. It is suitable for PAP detection system. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
HP-03 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3 | |
Horseradish peroxidase (Grade I). | |
0.1 mL | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction