Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HRP Antibody, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP305282
This item is not returnable.
View return policy
Description
HRP Polyclonal antibody specifically detects HRP in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
HRP | |
Polyclonal | |
Unconjugated | |
Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
Sheep | |
Delipidation and Defibrination | |
RUO | |
Reconstitute with 2.0 ml deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 1:1000-1:4000, ELISA 1:10000-1:40000, Immunohistochemistry, Immunoprecipitation | |
EC 1.11.1.7, Horseradish Peroxidase, HPRC 1, HPRC1, HRPA2, HRPC 2, HRPC2, HRPE5, HRPN, Neutral peroxidase, Peroxidase A2, Peroxidase C1A, Peroxidase C1B, Peroxidase C2, Peroxidase C3, Peroxidase E5, Peroxidase N, PRXC1A, PRXC1B, PRXC2, PRXC3 | |
HRP [Horseradish] (Uniprot: Q42517) | |
2 mL | |
Primary | |
Non-species specific |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction