Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP II Mouse, Clone: , Abnova™

Catalog No. 89934300 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-300 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-300 Supplier Abnova Corporation Supplier No. MAB12433
Add to Cart
Add to Cart

Mouse IgM monoclonal antibody raised against Plasmodium falciparum HRP II.

Specifications

Antigen HRP II
Applications Dot Blot, ELISA, Lateral Flow, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse IgM monoclonal antibody raised against Plasmodium falciparum HRP II.
Dilution Dot Blot ELISA Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Plasmodium falciparum HRP II
Purification Method Antigen Affinity Purification
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Parasite
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.