Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRP Monoclonal Antibody (3A5C6)
GREENER_CHOICE

Catalog No. PIMA515367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515367 Supplier Invitrogen™ Supplier No. MA515367
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15367 targets HRP in indirect ELISA, WB applications and shows reactivity with Tag samples. The MA5-15367 immunogen is purified recombinant fragment of HRP expressed in E. Coli.

HRP, also known as horseradish peroxidase, is an enzyme that is commonly used in biochemistry and molecular biology research. It is derived from the root of the horseradish plant and catalyzes the oxidation of a wide range of substrates in the presence of hydrogen peroxide. HRP is widely used as a detection enzyme in immunoassays, such as ELISAs and Western blots, due to its high sensitivity and stability. It can also be used as a tracer in enzyme-linked immunosorbent assays (ELISAs) and as a labeling agent in fluorescent microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A5C6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Alias Horseradish Peroxidase
Host Species Mouse
Immunogen Purified recombinant fragment of HRP expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.