Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRPT2/CDC73 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B4H2, Proteintech
SDP

Catalog No. 501736913 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6913 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6913 Supplier Proteintech Group Inc Supplier No. 664901IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.

Specifications

Antigen HRPT2/CDC73
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B4H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CDC73
Gene Accession No. Q4V8C8, Q6P1J9, Q8JZM7
Gene Alias C1orf28, CDC73, HPT JT, HRPT2, Hyperparathyroidism 2 protein, Parafibromin
Gene Symbols CDC73
Host Species Mouse
Immunogen HRPT2; CDC73 Fusion Protein ag2959
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100526068, 214498, 304832, 79577
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.