Learn More
HRS Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300988AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Involved in intracellular signal transduction mediated by cytokines and growth factors. When associated with STAM, it suppresses DNA signaling upon stimulation by IL-2 and GM-CSF. Could be a direct effector of PI3-kinase in vesicular pathway via early endosomes and may regulate trafficking to early and late endosomes by recruiting clathrin.Specifications
| HRS | |
| Polyclonal | |
| Unconjugated | |
| HGS | |
| HRS; Hepatocyte growth factor-regulated tyrosine kinase substrate; Protein pp110 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9146 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O14964 | |
| HGS | |
| Between 350 and 400 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.