Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HRV3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1118
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1118 Supplier Invitrogen™ Supplier No. PA1118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1118 has been used successfully in Western Blot to detect HRV3C cleavage site (Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro). PA1118 has been used in Immunoprecipitation of an HRV3C cleavage site-containing protein from a whole Hela cell lysate.

Human rhinovirus 3C protease (HRV3C Protease) is a cysteine protease that recognizes the cleavage site of Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro. It cleaves between Gln and Gly (independent of Pro). (HRV) 3C Protease is used to remove fusion tags from proteins with the HRV 3C cleavage sequence and is typically dual tagged for easy removal from the sample after cleavage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRV3C
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Alias HRV3; HRV-3; HRV-3C
Host Species Rabbit
Immunogen Synthetic peptide recognizing the cleavage site of HRV3c protease.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.