Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HRX Recombinant Rabbit Monoclonal Antibody (BL-175-7E8)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | HRX |
---|---|
Clone | BL-175-7E8 |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544215
|
Invitrogen™
MA544215 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544216
|
Invitrogen™
MA544216 |
20 μL |
Each for $173.10
|
|
|||||
Description
HRX Recombinant Monoclonal Antibody for Western Blot, IP
KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.Specifications
HRX | |
1 mg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
Q03164 | |
4297 | |
Synthetic peptide representing a region between residues 720 and 780 of human myeloid/lymphoid or mixed-lineage leukemia 1. | |
Antibody |
BL-175-7E8 | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
KMT2A | |
6430520K01; ALL1; ALL-1; CDK6/MLL fusion protein; C-terminal cleavage product of 180 kDa; CXXC7; CXXC-type zinc finger protein 7; histone-lysine N-methyltransferase 2A; histone-lysine N-methyltransferase MLL; HRX; HTRX; HTRX1; Kmt2a; lysine (K)-specific methyltransferase 2A; lysine methyltransferase 2A; lysine N-methyltransferase 2A; mixed lineage leukemia 1; mKIAA4050; Mll; MLL cleavage product C180; MLL cleavage product N320; MLL/ENL fusion protein; MLL/GAS7; MLL/GAS7 fusion protein; MLL/GMPS fusion protein; MLL/hCDCrel fusion protein; MLL1; MLL1A; MLL-AF4 der(11) fusion protein; MLL-AF9; Myeloid/lymphoid or mixed-lineage leukemia; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia 1; myeloid/lymphoid or mixed-lineage leukemia protein 1; N-terminal cleavage product of 320 kDa; p180; p320; rearranged MLL protein; TET1-MLL; trithorax Drosophila; trithorax-like protein; TRX1; WDSTS; zinc finger protein HRX | |
KMT2A | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title