Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HS1BP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576359
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene shares similarity with mouse Hs1bp3, an Hcls1/Hs1-interacting protein that may be involved in lymphocyte activation.
Specifications
HS1BP3 | |
Polyclonal | |
Unconjugated | |
Hs1bp3 | |
ETM2; HCLS1 binding protein 3; HCLS1-binding protein 3; HS1 binding protein 3; HS1-binding protein 3; Hs1bp3; HS1-BP3; HSP1BP-3; RGD1311331 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313950, 58240, 64342 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3TC93, Q53T59 | |
Hs1bp3 | |
Recombinant fusion protein of human HS1BP3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction