Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS1BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596983 Supplier Invitrogen™ Supplier No. PA596983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQSPAVLVTS RRLQNAHTGL DLTVPQHQEV RGKMMSGHVE YQILVVTRLA AFKSAKHRPE DVVQFLVSKK YSEIEEFYQK LSSRYAAASL PPLPRKVLFV GESDIRERRA VFNEILRCVS KDAELAGSPE LLEFLGTRSP GAAGLTSRDS SVLDGTDSQT GNDEEAFDFF EEQDQVAEEG PPVQSLKGED AEESLEEEEA; Positive Samples: HepG2, Mouse liver, Mouse kidney, Mouse heart, Rat liver.

The protein encoded by this gene shares similarity with mouse Hs1bp3, an Hcls1/Hs1-interacting protein that may be involved in lymphocyte activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS1BP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hs1bp3
Gene Accession No. Q3TC93, Q53T59
Gene Alias ETM2; HCLS1 binding protein 3; HCLS1-binding protein 3; HS1 binding protein 3; HS1-binding protein 3; Hs1bp3; HS1-BP3; HSP1BP-3; RGD1311331
Gene Symbols Hs1bp3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human HS1BP3 (NP_0719053).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313950, 58240, 64342
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.