Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS2ST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567369 Supplier Invitrogen™ Supplier No. PA567369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GATELYRTGKK SHLRKTTEKK LPTKQTIAKL QQSDIWKMEN EFYEFALEQF QFIRAHAVRE KDGDLYILAQ Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. This gene encodes a member of the heparan sulfate biosynthetic enzyme family that transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of this gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation. Two alternatively spliced transcript variants that encode different proteins have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS2ST1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HS2ST1
Gene Accession No. Q7LGA3
Gene Alias 2OST; 2-OST; 2-O-sulfotransferase; AW214369; dJ604K5.2; heparan sulfate 2-O-sulfotransferase 1; Hs2st; Hs2st1; KIAA0448; mKIAA0448
Gene Symbols HS2ST1
Host Species Rabbit
Immunogen Recombinant Human HS2ST1. Recombinant protein control fragment (Product #RP-108026).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9653
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.