Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS3ST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553630 Supplier Invitrogen™ Supplier No. PA553630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APRCLRGPSA GGQKLLQKSR PCDPSGPTPS EPSAPSAPAA AVPAPRLSGS NHSGSPKLGT KRLPQALIVG VKKGGTRAVL EFIRVHPDVR ALGTEPHFFD RNYGRG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. This gene is expressed predominantly in brain and may play a role in the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS3ST2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HS3ST2
Gene Accession No. Q9Y278
Gene Alias 30ST2; 3OST2; 3-OST-2; h3-OST-2; heparan sulfate (glucosamine) 3-O-sulfotransferase 2; heparan sulfate 3-O-sulfotransferase 2; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2; Heparan sulfate glucosamine 3-O-sulfotransferase 2; heparan sulfate-glucosamine 3-sulfotransferase 2; heparin-glucosamine 3-O-sulfotransferase; HS3ST2; UNQ2442/PRO5004
Gene Symbols HS3ST2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HS3ST2. Recombinant protein control fragment (Product #RP-90167).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9956
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.