Learn More
Invitrogen™ HS3ST2 Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | HS3ST2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HS3ST2 |
| Gene Accession No. | Q9Y278 |
| Gene Alias | 30ST2; 3OST2; 3-OST-2; h3-OST-2; heparan sulfate (glucosamine) 3-O-sulfotransferase 2; heparan sulfate 3-O-sulfotransferase 2; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2; Heparan sulfate glucosamine 3-O-sulfotransferase 2; heparan sulfate-glucosamine 3-sulfotransferase 2; heparin-glucosamine 3-O-sulfotransferase; HS3ST2; UNQ2442/PRO5004 |
| Gene Symbols | HS3ST2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.