Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS3ST3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566502 Supplier Invitrogen™ Supplier No. PA566502
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELAVWPAAAQ RKRLLQLPQW RRRRPPAPRD DGEEAAWEEE S Highest antigen sequence identity to the following orthologs - mouse 57%, rat 50%.

Sulfotransferase that utilizes 3'-phospho-5'-adenylyl sulfate (PAPS) to catalyze the transfer of a sulfo group to an N-unsubstituted glucosamine linked to a 2-O-sulfo iduronic acid unit on heparan sulfate. Catalyzes the O-sulfation of glucosamine in IdoUA2S-GlcNS and also in IdoUA2S-GlcNH2. The substrate-specific O-sulfation generates an enzyme-modified heparan sulfate which acts as a binding receptor to Herpes simplex virus-1 (HSV-1) and permits its entry. Unlike 3-OST-1, does not convert non-anticoagulant heparan sulfate to anticoagulant heparan sulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS3ST3A1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HS3ST3A1
Gene Accession No. Q9Y663
Gene Alias 30ST3A1; 3-OST-3A; 3OST3A1; h3-OST-3A; heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1; heparan sulfate 3-O-sulfotransferase 3A1; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1; Heparan sulfate glucosamine 3-O-sulfotransferase 3A1; heparan sulfate-glucosamine 3-sulfotransferase 3A1; heparin-glucosamine 3-O-sulfotransferase; HS3ST3A; HS3ST3A1; UNQ2551/PRO6180
Gene Symbols HS3ST3A1
Host Species Rabbit
Immunogen Recombinant Human HS3ST3A1. Recombinant protein control fragment (Product #RP-103795).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.