Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HS3ST3B1 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547910
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) HS3ST4, rhHS2ST1, rhHS6ST1, and recombinant mouse HS6ST3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
HS3ST3B1 is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta.Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta.
Specifications
HS3ST3B1 | |
Polyclonal | |
Unconjugated | |
HS3ST3B1 | |
30ST3B1; 3-OST-3B; 3OST3B1; h3-OST-3B; heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1; heparan sulfate 3-O-sulfotransferase 3B1; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3B1; Heparan sulfate glucosamine 3-O-sulfotransferase 3B1; heparan sulfate-glucosamine 3-sulfotransferase 3B1; HS3ST3B; HS3ST3B1; LOC100724084 | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
9953 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9Y662 | |
HS3ST3B1 | |
CHO-derived human Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Gly60-Asp390. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction