Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS3ST4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547809
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547809 Supplier Invitrogen™ Supplier No. PA547809
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HS3ST4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene HS3ST4
Gene Accession No. Q9Y661
Gene Alias 30ST4; 3OST4; 3-OST-4; EC 2.8.2; EC 2.8.2.23; EG628779; Gm6915; h3-OST-4; heparan sulfate (glucosamine) 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase-4; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4; Heparan sulfate glucosamine 3-O-sulfotransferase 4; heparan sulfate-glucosamine 3-sulfotransferase 4; Hs3st4
Gene Symbols HS3ST4
Host Species Sheep
Immunogen E. coli-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase4/HS3ST4 Gly184-Lys456.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 628779, 9951
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.