Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS3ST5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543288 Supplier Invitrogen™ Supplier No. PA543288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SQYNLYFNAT RGFYCLRFNI IFNKCLAGSK GRIHPEVDPS VITKLRKFFH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

HS3ST5 belongs to a group of heparan sulfate 3-O-sulfotransferases (EC 2.8.2.23) that transfer sulfate from 3-prime-phosphoadenosine 5-prime phosphosulfate (PAPS) to heparan sulfate and heparin (Mochizuki et al., 2003 [PubMed 12740361]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS3ST5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HS3ST5
Gene Accession No. Q8IZT8
Gene Alias 3OST5; 3-OST-5; D930005L05Rik; Gm1151; h3-OST-5; heparan sulfate (glucosamine) 3-O-sulfotransferase 5; heparan sulfate (glucosamine) 3-O-sulfotransferase 6; heparan sulfate 3-OST-5; heparan sulfate 3-O-sulfotransferase 5; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 5; Heparan sulfate glucosamine 3-O-sulfotransferase 5; heparan sulfate-glucosamine 3-sulfotransferase 5; Hs3ost5; HS3ST5; NBLA04021
Gene Symbols HS3ST5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human HS3ST5 (aa 280-329).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222537
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.