Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HS6ST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583238 Supplier Invitrogen™ Supplier No. PA583238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQLLRLQAFS SPVPDPYRSE DESSARFVPR YNFTRGDLLR KVDFDIKGDD LIVFLHIQK Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Heparan sulfate proteoglycans are ubiquitous components of the cell surface, extracellular matrix, and basement membranes, and interact with various ligands to influence cell growth, differentiation, adhesion, and migration. This gene encodes a member of the heparan sulfate (HS) sulfotransferase gene family, which catalyze the transfer of sulfate to HS. Different family members and isoforms are thought to synthesize heparan sulfates with tissue-specific structures and functions. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HS6ST2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HS6ST2
Gene Accession No. Q96MM7
Gene Alias 6OST2; EC-1; heparan sulfate 6-O-sulfotransferase 2; heparan-sulfate 6-O-sulfotransferase 2; Hs6st2; HS6ST-2; LOW QUALITY PROTEIN: heparan-sulfate 6-O-sulfotransferase 2; PSEC0092; RGD1563872; RGD1564397
Gene Symbols HS6ST2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HS6ST2. Recombinant protein control fragment (Product #RP-104647).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.