Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hs6st3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569568 Supplier Invitrogen™ Supplier No. PA569568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Human: 100%; Mouse: 93%; Pig: 93%.

6-O-sulfation enzyme which catalyzes the transfer of sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to position 6 of the N-sulfoglucosamine residue (GlcNS) of heparan sulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hs6st3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hs6st3
Gene Accession No. Q8IZP7
Gene Alias 6OST3; 6-ost-3; DKFZp761K2315; heparan sulfate 6-O-sulfotransferase 3; heparan sulfate 6-O-sulfotransferase 3b; heparan-sulfate 6-O-sulfotransferase 3; heparan-sulfate 6-O-sulfotransferase 3 pseudogene; heparan-sulfate 6-O-sulfotransferase 3-B; HS 6-OST-3-B; hs6st3; HS6ST-3; hs6st3b; RGD1560050; sb:eu962
Gene Symbols Hs6st3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human Hs6st3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266722
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.