Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103415 Supplier Invitrogen™ Supplier No. PA5103415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HSBP1.

Negative regulator of the heat shock response. Negatively affects HSF1 DNA-binding activity. May have a role in the suppression of the activation of the stress response during the aging process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSBP1
Gene Accession No. O75506, Q8K3X8, Q9CQZ1
Gene Alias 0610007A03Rik; AA408769; heat shock factor binding protein 1; heat shock factor-binding protein 1; HSBP1; HSF1BP; Hsp25; Nasopharyngeal carcinoma-associated antigen 13; NPC-A-13; Unknown (protein for MGC:128054)
Gene Symbols HSBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSBP1(Accession O75506), corresponding to amino acid residues L20-I70.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286899, 3281, 68196
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.