Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568701
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 86%.
Negative regulator of the heat shock response. Negatively affects HSF1 DNA-binding activity. May have a role in the suppression of the activation of the stress response during the aging process.
Specifications
HSBP1 | |
Polyclonal | |
Unconjugated | |
HSBP1 | |
0610007A03Rik; AA408769; heat shock factor binding protein 1; heat shock factor-binding protein 1; HSBP1; HSF1BP; Hsp25; Nasopharyngeal carcinoma-associated antigen 13; NPC-A-13; Unknown (protein for MGC:128054) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3281 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75506 | |
HSBP1 | |
synthetic peptide directed towards the N terminal region of human HSBP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction