Learn More
Hsc70 Mouse anti-Human, Mouse, Rat, Clone: 1A4B3, Proteintech
Shop All Proteintech Group Inc Products

Description
Specifications
Specifications
| Antigen | Hsc70 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1A4B3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HSPA8 |
| Gene Accession No. | P11142, P63017, P63018 |
| Gene Alias | Heat shock 70 kDa protein 8, heat shock 70kDa protein 8, HSC54, Hsc70, HSC71, HSP71, HSP73, HSPA10, HSPA8, LAP1, NIP71 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.