Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSC70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518537
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518537 Supplier Invitrogen™ Supplier No. PA518537
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. HSC70 (also known as HSC71, HSC73, HSP73, p72, prp73) is expressed constitutively and is slightly heat-inducible. HSC70 binds to the exposed loop of clathrin light chains to promote uncoating and can also bind the cytoskeleton which may facilitate cytoskeletal rearrangements. HSC70 has been shown to stimulate lysosomal degradation of intracellular proteins and to retard both aggregation and folding of mitochondrial precursor proteins in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSC70
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Hspa8
Gene Accession No. P11142, P63017
Gene Alias 2410008N15Rik; AAG2; constitutive heat shock protein 70; constitutively expressed heat shock protein; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; Heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 8; heat shock 70kDa protein 8; heat shock cognate; heat shock cognate 70; heat shock cognate 71 kD protein; heat shock cognate 71 kDa protein; heat shock cognate 71 kDa protein-like protein; heat shock cognate 71-kd protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; heat shock protein family A (Hsp70) member 8; HEL-33; HEL-S-72p; HIP; HSC54; Hsc70; Hsc70 ATPase; HSC70 protein; Hsc71; Hsc73; HSP70; HSP71; HSP73; Hspa10; Hspa8; hspa8 protein; I79_022974; LAP; LAP1; LAP-1; lipopolysaccharide-associated protein 1; LPS-associated protein 1; MGC131511; MGC29929; NIP71; N-myristoyltransferase inhibitor protein 71; Protein FAM10A1; similar to 7O KDa heat shock cognate protein; SNC6; unnamed protein product; wu:fb01g06; wu:fi48b06
Gene Symbols Hspa8
Host Species Goat
Immunogen Synthetic peptide sequence (EKLQGKINDEDKQK) corresponding to the internal amino acids of HSPA8 (aa 556-569).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15481, 3312
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.