Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSCB Antibody (OTI3E1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272415V
Description
HSCB Monoclonal specifically detects HSCB in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HSCB | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSCB | |
Full length recombinant protein of human HSCB (NP_741999) produced in HEK293T cells | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3E1 | |
Western Blot, Immunohistochemistry | |
DnaJ homolog subfamily C member 20, DNAJC20DnaJ (Hsp40) homolog, subfamily C, member 20, Hsc20, HSC20dJ366L4.2, HscB iron-sulfur cluster co-chaperone homolog (E. coli), iron-sulfur cluster co-chaperone protein HscB, mitochondrial, Jac1, J-type co-chaperone HSC20, MGC2637, MGC74462 | |
Mouse | |
Affinity Purified | |
metabolism | |
150274.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction