Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598609
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598609 Supplier Invitrogen™ Supplier No. PA598609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSCB Polyclonal Antibody for Western Blot, IHC (P), ELISA

May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSCB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.855 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HSCB
Gene Accession No. Q8IWL3
Gene Alias AI325508; AW049829; C-HSC20; dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; DNAJC20; HSC20; HSCB; HscB iron-sulfur cluster co-chaperone homolog; HscB iron-sulfur cluster co-chaperone homolog (E. coli); HscB mitochondrial iron-sulfur cluster cochaperone; HscB mitochondrial iron-sulfur cluster co-chaperone; iron-sulfur cluster co-chaperone protein HscB; Iron-sulfur cluster co-chaperone protein HscB, cytoplasmic; Iron-sulfur cluster co-chaperone protein HscB, mitochondrial; JAC1; J-type co-chaperone HSC20; RGD1311005
Gene Symbols HSCB
Host Species Rabbit
Immunogen Recombinant Human Iron-sulfur cluster co-chaperone protein HscB, mitochondrial protein (30-235AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150274, 360826
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.